Thiamin Hydrochlorid 1 g/ml in Wasser

Lieferant: PanReac AppliChem

Synonyme: Thiaminiumchlorid, Antiberiberifaktor Hydrochlorid, antineuritisches Vitamin Hydrochlorid, Aneurin Hydrochlorid, Thiaminchlorid Hydrochlorid, Vitamin B1 Hydrochlorid

A8094.1000
APPCA8094.1000EA 0 EUR
APPCA8094.1000
Thiamin Hydrochlorid 1 g/ml in Wasser
Thiamin Hydrochlorid
Formel: C₁₂H₁₈Cl₂N₄OS
Lagertemperatur: Room Temperature
MDL: MFCD00012780
CAS-Nummer: 67-03-8
EINECS: 200-641-8

Bestellen

Spezifikation Testergebnisse

Assay (titr., calc. on dried subst.) 98.5 - 101.0%
Appearance of solution Passes test
Identity Passes test
pH (2.5 %; H2O) 2.7 - 3.3
Water (K.F.) max. 5 %
Sulfated ash max. 0.1 %
Sulfate max. 0.03 %
Related subst. (HPLC) Impurity B: max. 0.3 %
Impurity A: max. 0.15 %
Impurity C: max. 0.15 %
Each unspecified impurity max. 0.10 %
Total impurities max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR