VWR® RediSep® RF, Disposable columns
Lieferant: VWR Chemicals
AS-69-2203-380EA 578 EUR
AS-69-2203-380
VWR® RediSep® RF, Disposable columns
Chromatographiesäulen
RediSep® Rf disposable, bare silica columns for flash purification of organic compounds are the benchmark for normal phase separations. This high quality media is specified for high resolution and reproducibility for everyday purifications.
Loading volume: 0,1 to 10%