3-Chlorisonicotinonitril

Lieferant: ASTATECH

31020-5G
ASTA31020-5GEA 0 EUR
ASTA31020-5G
3-Chlorisonicotinonitril
3-Chlorisonicotinonitril
Formel: C₆H₃ClN₂
Molecular Weight: 138.56 g/mol
MDL: MFCD05663706
CAS-Nummer: 68325-15-5
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR