3-Formyl-6-methoxypyridin

Lieferant: ASTATECH

Synonyme: 6-Methoxypyridine-3-carbaldehyde, 6-Methoxynicotinaldehyde, 6-Methoxypyridine-3-carboxaldehyde, 2-Methoxypyridine-5-carboxaldehyde, 6-Methoxy-3-pyridinecarboxaldehyde

64881
ASTA64881EA 0 EUR
ASTA64881
3-Formyl-6-methoxypyridin
3-Formyl-6-methoxypyridin
Formel: C₇H₇NO₂
Molecular Weight: 137,14 g/mol
MDL: MFCD02683446
CAS-Nummer: 65873-72-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR