4'-Brom-3'-chloracetophenon

Lieferant: ASTATECH

Synonyme: 1-(4-Bromo-3-chlorophenyl)ethanone

CL8916-5G
ASTACL8916-5GEA 0 EUR
ASTACL8916-5G
4'-Brom-3'-chloracetophenon
4'-Brom-3'-chloracetophenon
Formel: C₈H₆BrClO
Molecular Weight: 233,4900 g/mol
MDL: MFCD14636445
CAS-Nummer: 3114-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR