Anti-GRIK2 Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
ARP35340_T100
AVIVARP35340_T100EA 349 EUR
AVIVARP35340_T100
Anti-GRIK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GRIK2 (glutamate receptor, ionotropic, kainate 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR