Anti-IBSP Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP36681_T100
AVIVARP36681_T100EA 349 EUR
AVIVARP36681_T100
Anti-IBSP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IBSP (integrin-binding sialoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human