Anti-SLC1A1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP43865_P050
AVIVARP43865_P050EA 401 EUR
AVIVARP43865_P050
Anti-SLC1A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC1A1 (solute carrier family 1 (neuronal/epithelial high affinity glutamate transporter, system Xag), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen