Anti-MINOS1 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP44801_P050
AVIVARP44801_P050EA 401 EUR
AVIVARP44801_P050
Anti-MINOS1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MINOS1 (mitochondrial inner membrane organizing system 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Bovine, Dog, Human, Mouse, Rat