Anti-STAC2 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP69675_P050
AVIVARP69675_P050EA 401 EUR
AVIVARP69675_P050
Anti-STAC2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: STAC2 (SH3 and cysteine rich domain 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human