Anti-BPIFB3 Rabbit Polyclonal Antibody
Lieferant: Aviva Systems Biology
ARP69708_P050
AVIVARP69708_P050EA 401 EUR
AVIVARP69708_P050
Anti-BPIFB3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: BPIFB3 (BPI fold containing family B, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human