Anti-GABRD Rabbit Polyclonal Antibody

Lieferant: Aviva Systems Biology
AVARP13032P050
AVIVAVARP13032P050EA 401 EUR
AVIVAVARP13032P050
Anti-GABRD Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GABRD (gamma-aminobutyric acid (GABA) A receptor, delta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR