Microcentrifuge tube rack, Axygen®
Lieferant: Corning
TR8210
AXYGTR8210EA 58.9 EUR
AXYGTR8210
Microcentrifuge tube rack, Axygen®
Gestelle Reagenzglasgestelle
Microcentrifuge tube rack is molded from durable, autoclavable polypropylene, and features non-skid rubber feet.
  • Step design allows easy access to tubes
  • Two rows of 10 (20×1,5 to 2,0 ml total)
The rack has two levels and exhibits wide spacing to provide prime sample access and visibility. Tube positions are numbered 1 to 20 for organizing samples.
Verpackung: Two in a case.