Jerricans, narrow mouth, HDPE, Azlon®
Lieferant: SciLabware
JGH140 JGH148
AZLOJGH140EA 13.3 EUR
AZLOJGH140 AZLOJGH148
Jerricans, narrow mouth, HDPE, Azlon®
Flaschen Kanister
Plastic, translucent.
  • Lightweight and robust
  • Corrosion resistant
  • Ideal for storage and transportation of liquid chemicals
  • Suitable for food contact applications in accordance with EU and U.S. FDA regulations