Water jet pump
Lieferant: SciLabware
PWD003
AZLOPWD003EA 76.5 EUR
AZLOPWD003
Water jet pump
Pumpen Wasserstrahl-Vakuumpumpen
Water jet pump suitable for most laboratory water pressures with integrated non-return valve that increases safety and a detachable vacuum connection for 6 to 9 mm bore tubing.
Lieferumfang: Supplied with water inlet hose connection and R ³/₄" threaded tap connectors, with R ¹/₂" reduction adapter.