Holder for round bottom flasks
Lieferant: SciLabware
TWR038
AZLOTWR038EA 41.7 EUR
AZLOTWR038
Holder for round bottom flasks
Halterungen und Träger Kolben- und Becherhalterungen
This stand is suitable for most round bottomed flasks.
  • Good chemical resistance
  • Stepped concentric rings to help stabilise flask