Bathocuproinsulfonat Dinatriumsalz Hydrat determination_Cu_Fe_ spectrophotometric, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Bathocuproindisulphonic disodium salt hydrate
MQ200
B1125-1G
B1125-25G
B1125-500MG
B1125-5G
B1125-1GEA
180
EUR
B1125-1G
B1125-25G
B1125-500MG
B1125-5G
Bathocuproinsulfonat Dinatriumsalz Hydrat determination_Cu_Fe_ spectrophotometric, Sigma-Aldrich®
Bathocuproinsulfonat Dinatriumsalz Hydrat
Formel:
C₂₆H₁₈N₂Na₂O₆S₂·H₂O Molecular Weight: 564.55 g/mol (anhy) Lagertemperatur: Room Temperature |
MDL:
MFCD03012947 CAS-Nummer: 52698-84-7 EINECS: 258-111-7 |
|
Spezifikation Testergebnisse
Appearance (Colour) | White to Beige |
Appearance (Form) | Powder or Crystals |
Titration with HClO₄ | ≥90% |
Solubility (Colour) | Colourless to Very Dark Yellow to Very Dark Brown and Colourless to Very Dark Red to Very Dark Purple |
Solubility (Turbidity), 50 mg/ml in Water | Clear to Hazy |
Water | ≤12.0% |
Infrared Spectrum | Conforms to Structure |
Application Test, Suitable for Use in the Determination of Copper | Corresponds to Requirements |
Assay (ICP) | 7.2 - 9.1% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...