Bathocuproinsulfonat Dinatriumsalz Hydrat determination_Cu_Fe_ spectrophotometric, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Bathocuproindisulphonic disodium salt hydrate

MQ200

B1125-1G B1125-25G B1125-500MG B1125-5G
B1125-1GEA 180 EUR
B1125-1G B1125-25G B1125-500MG B1125-5G
Bathocuproinsulfonat Dinatriumsalz Hydrat determination_Cu_Fe_ spectrophotometric, Sigma-Aldrich®
Bathocuproinsulfonat Dinatriumsalz Hydrat
Formel: C₂₆H₁₈N₂Na₂O₆S₂·H₂O
Molecular Weight: 564.55 g/mol (anhy)
Lagertemperatur: Room Temperature
MDL: MFCD03012947
CAS-Nummer: 52698-84-7
EINECS: 258-111-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98c0ab448486c4c2ab1f4826f9c4ac422c581065&vid=625c7c42d2d81ce70aebb4abfab261eb82483afb&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Beige
Appearance (Form) Powder or Crystals
Titration with HClO₄ ≥90%
Solubility (Colour) Colourless to Very Dark Yellow to Very Dark Brown and Colourless to Very Dark Red to Very Dark Purple
Solubility (Turbidity), 50 mg/ml in Water Clear to Hazy
Water ≤12.0%
Infrared Spectrum Conforms to Structure
Application Test, Suitable for Use in the Determination of Copper Corresponds to Requirements
Assay (ICP) 7.2 - 9.1%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR