Bathophenanthrolindisulfonsäure Dinatriumsalz Hydrat 95, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
4,7-Diphenyl-1,10-phenanthrolinedisulphonic acid disodium salt hydrate
MQ200
Bathophenanthrolinedisulfonic acid disodium salt hydrate has been used to measure iron to determine the solubility of iron in the biological sample. It has been used as an iron chelator to grow plants without Fe-EDTA (ethylenediaminetetraacetic acid).
Bathophenanthrolinedisulfonic acid is a metal chelator. It permits iron determination to be made in aqueous medium without need for solvent extraction. It forms a complex with Fe2+ and makes it unavailable to Fe2+ transporter, thereby inhibiting the transport of iron into chloroplasts. Bathophenanthrolinedisulfonic acid is also used in the assay for determining intracellular glutathione.
Formel:
C₂₄H₁₄N₂Na₂O₆S₂·H₂O Molecular Weight: 536.48 g/mol (anhy) Schmelzpunkt: 300 °C (Lit.) Lagertemperatur: Room Temperature |
MDL:
MFCD00149310 CAS-Nummer: 52746-49-3 EINECS: 258-152-0 |
|
Spezifikation Testergebnisse
Appearance (colour) | White to Orange and Faint Beige to Beige and Pink |
Appearance (form) | Powder |
Titration (NT) HCLO₄ 0.1M | calculated on dry substance |
Purity (HPLC area %) | ≥ 94.5 % |
Solubility (turbidity) | Clear to Slightly Hazy |
Solubility (method) | 100 mg/ml in water |
Water | Report Result |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...