Methyl-3,4-dichlorbenzoat ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl 3,4-dichlorobenzoate, Methyl 3,4-dichlorobenzenecarboxylate

B20027.06
B20027.06EA 67.5 EUR
B20027.06
Methyl-3,4-dichlorbenzoat ≥97%
Methyl-3,4-dichlorbenzoat

Purity: 97% CAS No.: 2905-68-2 Molecular Formula: C8H6Cl2O2 Formula Weight: 205.04 Melting Point: 44-46° MDL No.: MFCD00018165

Formel: C₈H₆Cl₂O₂
Molecular Weight: 205,04 g/mol
Schmelzpunkt: 44…46 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00018165
CAS-Nummer: 2905-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR