Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Dimethyl 4-aminothiophene-2,3-dicarboxylate hydrochloride
B20028.03
B20028.03EA
67.9
EUR
B20028.03
Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride 97%
Dimethyl-4-aminothiophene-2,3-dicarboxylate hydrochloride
Purity: 97%
CAS No.: 121071-71-4
Molecular Formula: C8H9NO4S.HCl
Formula Weight: 251.69
Melting Point: 166-169°
MDL No.: MFCD00205189
Formel:
C₈H₁₀ClNO₄S Molecular Weight: 251.681 g/mol Schmelzpunkt: 166…169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00205189 CAS-Nummer: 121071-71-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...