4-(Trifluormethoxy)phenylisocyanat ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 35037-73-1
Molecular Formula: C8H4F3NO2
Formula Weight: 203.12
Storage Temperature: 2°C to 8°C
Flash Point: 70°(158°F)
Refractive Index: 1.4580
Melting Point: -29°
Boiling Point: 78°/22mm
Beilstein Reference: 977669
UN No.: 2206
MDL No.: MFCD00035702
Formel:
C₈H₄F₃NO₂ Siedepunkt: 78 °C (22 mmHg) Schmelzpunkt: –29 °C Dichte: 1,346 Flammpunkt: 70 °C (158 °F) |
MDL:
MFCD00035702 CAS-Nummer: 35037-73-1 EINECS: 252-328-0 UN: 2206 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...