6-Hydroxy-m-toluylsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Methylsalicylic acid, 2,5-Cresotic acid

B20040.06 B20040.14
B20040.06EA 41.9 EUR
B20040.06 B20040.14
6-Hydroxy-m-toluylsäure ≥98%
6-Hydroxy-m-toluylsäure

Purity: 98% CAS No.: 89-56-5 Molecular Formula: C8H8O3 Formula Weight: 152.15 Odour: Odourless Melting Point: 151-154° MDL No.: MFCD00002461

Formel: C₈H₈O₃
Molecular Weight: 152.15 g/mol
Siedepunkt: 313
Schmelzpunkt: 151…154 °C
Dichte: 1.31 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002461
CAS-Nummer: 89-56-5
EINECS: 201-918-6
Merck Index: 12,02650

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR