4-Brom-2-ethyl-1-iodbenzol ≥97% stabilisiert

Lieferant: Thermo Fisher Scientific
Achtung

B20052.09 B20052.88 B20052.36
B20052.09EA 166 EUR
B20052.09 B20052.88 B20052.36
4-Brom-2-ethyl-1-iodbenzol ≥97% stabilisiert
4-Brom-2-ethyl-1-iodbenzol

Stabilized with copper


Purity: 97% CAS No.: 175278-30-5 Molecular Formula: C8H8BrI Formula Weight: 310.97 Boiling Point: 84-86°/0.4mm MDL No.: MFCD00221458

Formel: C₈H₈BrI
Molecular Weight: 310.97 g/mol
Siedepunkt: 84…86 °C (0.4 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00221458
CAS-Nummer: 175278-30-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR