4-Brom-3-methyl-1H-pyrazol ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Bromo-3-methylpyrazole
B20069.06
B20069.06EA
156
EUR
B20069.06
4-Brom-3-methyl-1H-pyrazol ≥97%
4-Bromo-3-methyl-1H-pyrazole
Purity: 97%
CAS No.: 13808-64-5
Molecular Formula: C4H5BrN2
Formula Weight: 161.01
Odour: Odourless
Melting Point: 73-77°
MDL No.: MFCD00005241
Formel:
C₄H₅BrN₂ Molecular Weight: 161,01 g/mol Schmelzpunkt: 73…77 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005241 CAS-Nummer: 13808-64-5 EINECS: 237-456-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...