trans-Aconitsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B20087.14
B20087.22
B20087.PG
B20087.14EA
60.9
EUR
B20087.14
B20087.22
B20087.PG
trans-Aconitsäure ≥98%
trans-Aconitsäure
Purity: 98%
CAS No.: 4023-65-8
Molecular Formula: C6H6O6
Formula Weight: 174.11
Odour: Odourless
Melting Point: ca 190° dec.
MDL No.: MFCD00002721
Formel:
C₆H₆O₆ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002721 CAS-Nummer: 4023-65-8 EINECS: 223-688-6 Merck Index: 14,117 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...