1-Cyclohexyl-1-butanol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: α-Propylcyclohexanemethanol

B20090.14
B20090.14EA 115 EUR
B20090.14
1-Cyclohexyl-1-butanol ≥98%
1-Cyclohexyl-1-butanol

Purity: 98% CAS No.: 4352-42-5 Molecular Formula: C10H20O Formula Weight: 156.27 Flash Point: 108°(228°F) Refractive Index: 1.4650 Boiling Point: 103-104°/4mm MDL No.: MFCD00019360

Formel: C₁₀H₂₀O
Molecular Weight: 156.27 g/mol
Siedepunkt: 103…104 °C (4 mmHg)
Dichte: 0.902
Flammpunkt: 108 °C (228 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00019360
CAS-Nummer: 4352-42-5
EINECS: 224-419-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR