1-Brom-4-(difluormethoxy)benzol ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B20106.06 B20106.14
B20106.06EA 114 EUR
B20106.06 B20106.14
1-Brom-4-(difluormethoxy)benzol ≥97%
1-Brom-4-(difluormethoxy)benzol

Purity: 97% CAS No.: 5905-69-1 Molecular Formula: C7H5BrF2O Formula Weight: 223.02 Refractive Index: 1.5010 Boiling Point: 96-98°/15mm UN No.: 1993 MDL No.: MFCD00221465

Formel: C₇H₅BrF₂O
Siedepunkt: 96…98 °C (15 mmHg)
Dichte: 1,631
Lagertemperatur: Raumtemperatur
MDL: MFCD00221465
CAS-Nummer: 5905-69-1
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR