Tetrabromphenolblau

Lieferant: Thermo Fisher Scientific

Synonyme: 3,3',3',4,5,5',5′',6-Octabromphenolsulfonphthalein, 4,4'-(4,5,6,7-Tetrabrom-3H-2,1-benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid

B20123.03 B20123.06 B20123.14
B20123.03EA 34.9 EUR
B20123.03 B20123.06 B20123.14
Tetrabromphenolblau
Tetrabromphenolblau

CAS No.: 4430-25-5 Molecular Formula: C19H6Br8O5S Formula Weight: 985.55 Melting Point: 203-205° dec. Beilstein Reference: 378438 MDL No.: MFCD00005876

Formel: C₁₉H₆Br₈O₅S
Molecular Weight: 985.55 g/mol
Schmelzpunkt: 203…205 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005876
CAS-Nummer: 4430-25-5
EINECS: 224-622-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR