(±)-6-Hepten-3-ol ≥99%

Lieferant: Thermo Scientific
Achtung

B20128.03
B20128.03EA 25.6 EUR
B20128.03
(±)-6-Hepten-3-ol ≥99%
(±)-6-Hepten-3-ol

Purity: 99% CAS No.: 19781-77-2 Molecular Formula: C7H14O Formula Weight: 114.18 UN No.: 1987 MDL No.: MFCD00046683

Formel: C₇H₁₄O
Molecular Weight: 114,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00046683
CAS-Nummer: 19781-77-2
UN: 1987
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR