1H,1H,7H-Dodecafluor-1-heptanol ≥97%
Lieferant: Thermo Fisher Scientific
B20144.14
B20144.14EA
46
EUR
B20144.14
1H,1H,7H-Dodecafluor-1-heptanol ≥97%
1H,1H,7H-Dodecafluor-1-heptanol
Purity: 97%
CAS No.: 335-99-9
Molecular Formula: H(CF2)6CH2OH
Formula Weight: 332.09
Flash Point: None
Refractive Index: 1.3180
Melting Point: -20°
Boiling Point: 169-170°
Beilstein Reference: 1800110
MDL No.: MFCD00039630
Formel:
C₇H₄F₁₂O Siedepunkt: 169…170 °C Schmelzpunkt: –20 °C Dichte: 1,762 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039630 CAS-Nummer: 335-99-9 EINECS: 206-406-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...