(±)-1-Penten-3-ol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (±)-Ethylvinylcarbinol, (±)-Ethyl-vinylcarbinol

B20155.06
B20155.06EA 39.5 EUR
B20155.06
(±)-1-Penten-3-ol ≥98%
(±)-1-Penten-3-ol

Purity: 98+% CAS No.: 616-25-1 Molecular Formula: C5H10O Formula Weight: 86.13 Flash Point: 25°(77°F) Refractive Index: 1.4250 Boiling Point: 114-115° Beilstein Reference: 1719834 UN No.: 1987 MDL No.: MFCD00004573

Formel: C₅H₁₀O
Siedepunkt: 114…115 °C
Dichte: 0,838
Flammpunkt: 25 °C (77 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004573
CAS-Nummer: 616-25-1
EINECS: 210-472-1
UN: 1987
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR