1H,1H,2H,2H-Tridecafluor-1-octanol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B20156.18
B20156.09
B20156.30
B20156.18EA
194
EUR
B20156.18
B20156.09
B20156.30
1H,1H,2H,2H-Tridecafluor-1-octanol ≥97%
1H,1H,2H,2H-Tridecafluor-1-octanol
Purity: 97%
CAS No.: 647-42-7
Molecular Formula: CF3(CF2)5(CH2)2OH
Formula Weight: 364.11
Flash Point: 91°(195°F)
Refractive Index: 1.3140
Boiling Point: 90°/20mm
Beilstein Reference: 1992757
MDL No.: MFCD00042143
Formel:
C₈H₅F₁₃O Siedepunkt: 90 °C (20 mmHg) Dichte: 1,68 Flammpunkt: 91 °C (195 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042143 CAS-Nummer: 647-42-7 EINECS: 211-477-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...