N,N-Dimethylethylendiamin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

B20182.18 B20182.22 B20182.30
B20182.18EA 39.5 EUR
B20182.18 B20182.22 B20182.30
N,N-Dimethylethylendiamin 97%
N,N-Dimethylethylendiamin

Purity: 97% CAS No.: 108-00-9 Molecular Formula: C4H12N2 Formula Weight: 88.15 Flash Point: 12°(53°F) Refractive Index: 1.4270 Melting Point: -70° Boiling Point: 107° Beilstein Reference: 605279 UN No.: 2733 MDL No.: MFCD00008175

Formel: C₄H₁₂N₂
Siedepunkt: 107 °C
Schmelzpunkt: < –70 °C
Dichte: 0,810
Flammpunkt: 12 °C (53 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008175
CAS-Nummer: 108-00-9
EINECS: 203-541-2
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR