3-Nitro-o-phenylendiamin 98%

Lieferant: Thermo Fisher Scientific
Achtung

B20201.03 B20201.06 B20201.14
B20201.03EA 53.4 EUR
B20201.03 B20201.06 B20201.14
3-Nitro-o-phenylendiamin 98%
3-Nitro-o-phenylendiamin

Purity: 98% CAS No.: 3694-52-8 Molecular Formula: C6H7N3O2 Formula Weight: 153.14 Melting Point: 157-162° Beilstein Reference: 909344

Formel: C₆H₇N₃O₂
Schmelzpunkt: 157…162 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007722
CAS-Nummer: 3694-52-8
EINECS: 223-013-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR