1-Iodperfluoroctan ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B20207.09
B20207.09EA 46.9 EUR
B20207.09
1-Iodperfluoroctan ≥98%
1-Iodperfluoroctan

Purity: 98% CAS No.: 507-63-1 Molecular Formula: C8F17I Formula Weight: 545.97 Refractive Index: 1.3290 Melting Point: 25° Boiling Point: 160-161° Beilstein Reference: 1717141 MDL No.: MFCD00001064

Formel: C₈F₁₇I
Siedepunkt: 160…161 °C
Schmelzpunkt: 25 °C
Dichte: 2,040
Lagertemperatur: Raumtemperatur
MDL: MFCD00001064
CAS-Nummer: 507-63-1
EINECS: 208-079-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR