Methyl-(3-methoxyphenyl)acetat ≥97%
Lieferant: Thermo Fisher Scientific
Synonyme:
Methyl 3-Methoxyphenylacetate
B20237.09
B20237.18
B20237.88
B20237.09EA
141
EUR
B20237.09
B20237.18
B20237.88
Methyl-(3-methoxyphenyl)acetat ≥97%
Methyl-(3-methoxyphenyl)acetat
Purity: 97%
CAS No.: 18927-05-4
Molecular Formula: C10H12O3
Formula Weight: 180.23
Boiling Point: 86-88°/0.5mm
MDL No.: MFCD00017205
Formel:
C₁₀H₁₂O₃ Siedepunkt: 86…88 °C (0,5 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017205 CAS-Nummer: 18927-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...