Methyl-(3-methoxyphenyl)acetat ≥97%

Lieferant: Thermo Fisher Scientific

Synonyme: Methyl 3-Methoxyphenylacetate

B20237.09 B20237.18 B20237.88
B20237.09EA 141 EUR
B20237.09 B20237.18 B20237.88
Methyl-(3-methoxyphenyl)acetat ≥97%
Methyl-(3-methoxyphenyl)acetat

Purity: 97% CAS No.: 18927-05-4 Molecular Formula: C10H12O3 Formula Weight: 180.23 Boiling Point: 86-88°/0.5mm MDL No.: MFCD00017205

Formel: C₁₀H₁₂O₃
Siedepunkt: 86…88 °C (0,5 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00017205
CAS-Nummer: 18927-05-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR