2-Methoxyphenyl-acetat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
1-Acetoxy-2-methoxybenzene, Acetic acid 2-methoxyphenyl ester, 2-Acetoxyanisole, Acetic acid 2-guaiacol ester, 2-Guaiacol acetate, O-Acetylguaiacol
B20242.14
B20242.22
B20242.14EA
36.6
EUR
B20242.14
B20242.22
2-Methoxyphenyl-acetat ≥98%
2-Methoxyphenyl acetate
Purity: 98%
CAS No.: 15212-03-0
Molecular Formula: C9H10O3
Formula Weight: 166.18
MDL No.: MFCD00017221
Formel:
C₉H₁₀O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017221 CAS-Nummer: 15212-03-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...