2-Methoxyphenyl-acetat ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 1-Acetoxy-2-methoxybenzene, Acetic acid 2-methoxyphenyl ester, 2-Acetoxyanisole, Acetic acid 2-guaiacol ester, 2-Guaiacol acetate, O-Acetylguaiacol

B20242.14 B20242.22
B20242.14EA 36.6 EUR
B20242.14 B20242.22
2-Methoxyphenyl-acetat ≥98%
2-Methoxyphenyl acetate

Purity: 98% CAS No.: 15212-03-0 Molecular Formula: C9H10O3 Formula Weight: 166.18 MDL No.: MFCD00017221

Formel: C₉H₁₀O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00017221
CAS-Nummer: 15212-03-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR