3,4-Dimethyl-2-hexanol (Gemisch der Stereoisomere) erythro + threo ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B20243.03
B20243.14
B20243.03EA
32.9
EUR
B20243.03
B20243.14
3,4-Dimethyl-2-hexanol (Gemisch der Stereoisomere) erythro + threo ≥98%
3,4-Dimethyl-2-hexanol (Gemisch der Stereoisomere)
Purity: 98%
CAS No.: 31350-88-6
Molecular Formula: C8H18O
Formula Weight: 130.23
MDL No.: MFCD00021887
Formel:
C₈H₁₈O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021887 CAS-Nummer: 31350-88-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...