1,4-Bis(2-hydroxyhexafluorisopropyl)benzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,2'-(Benzene-1,4-diyl)bis(1,1,1,3,3,3-hexafluoropropan-2-ol)

B20259.09
B20259.09EA 182 EUR
B20259.09
1,4-Bis(2-hydroxyhexafluorisopropyl)benzol ≥98%
1,4-Bis(2-hydroxyhexafluorisopropyl)benzol

Purity: 98% CAS No.: 1992-15-0 Molecular Formula: C12H6F12O2 Formula Weight: 410.16 Melting Point: 92-95° Boiling Point: 100°/20mm MDL No.: MFCD00042091

Formel: C₁₂H₆F₁₂O₂
Molecular Weight: 410,16 g/mol
Siedepunkt: 100 °C (20 mmHg)
Schmelzpunkt: 92…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00042091
CAS-Nummer: 1992-15-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR