cis-3-Methylcyclohexanol 95%

Lieferant: Thermo Scientific

B20288.06
B20288.06EA 116 EUR
B20288.06
cis-3-Methylcyclohexanol 95%
cis-3-Methylcyclohexanol

Purity: 95% CAS No.: 24965-90-0 Molecular Formula: C7H14O Formula Weight: 114.19 Flash Point: 73°(163°F) MDL No.: MFCD00067155

Formel: C₇H₁₄O
Molecular Weight: 114,19 g/mol
Flammpunkt: 73 °C (163 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00067155
CAS-Nummer: 24965-90-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR