4-Morpholinoanilin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-(4-Morpholinyl)aniline

B20292.06 B20292.14
B20292.06EA 39.7 EUR
B20292.06 B20292.14
4-Morpholinoanilin ≥98%
4-Morpholinoanilin

Purity: 98+% CAS No.: 2524-67-6 Molecular Formula: C10H14N2O Formula Weight: 178.24 Melting Point: 129-133° Beilstein Reference: 150957 UN No.: 2811 MDL No.: MFCD00006169

Formel: C₁₀H₁₄N₂O
Molecular Weight: 178.23 g/mol
Schmelzpunkt: 129…133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006169
CAS-Nummer: 2524-67-6
EINECS: 219-760-1
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR