3-Ethoxy-2-thenoesäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Ethoxythiophene-2-carboxylic acid
B20296.03
B20296.06
B20296.14
B20296.03EA
66.9
EUR
B20296.03
B20296.06
B20296.14
3-Ethoxy-2-thenoesäure ≥97%
3-Ethoxy-2-thenoesäure
Purity: 97%
CAS No.: 139926-23-1
Molecular Formula: C7H8O3S
Formula Weight: 172.20
Melting Point: 121-123°
MDL No.: MFCD00205205
Formel:
C₇H₈O₃S Molecular Weight: 172.2 g/mol Schmelzpunkt: 121…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00205205 CAS-Nummer: 139926-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...