3-Ethoxy-2-thenoesäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Ethoxythiophene-2-carboxylic acid

B20296.03 B20296.06 B20296.14
B20296.03EA 66.9 EUR
B20296.03 B20296.06 B20296.14
3-Ethoxy-2-thenoesäure ≥97%
3-Ethoxy-2-thenoesäure

Purity: 97% CAS No.: 139926-23-1 Molecular Formula: C7H8O3S Formula Weight: 172.20 Melting Point: 121-123° MDL No.: MFCD00205205

Formel: C₇H₈O₃S
Molecular Weight: 172.2 g/mol
Schmelzpunkt: 121…123 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00205205
CAS-Nummer: 139926-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR