5-Bromindol ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Bromo-1H-indole
B20307.06
B20307.14
B20307.22
B20307.06EA
29.3
EUR
B20307.06
B20307.14
B20307.22
5-Bromindol ≥99%
5-Bromindol
Purity: 99%
CAS No.: 10075-50-0
Molecular Formula: C8H6BrN
Formula Weight: 196.05
Odour: Odourless
Melting Point: 89-92°
Beilstein Reference: 112877
MDL No.: MFCD00005670
Formel:
C₈H₆BrN Schmelzpunkt: 89…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005670 CAS-Nummer: 10075-50-0 EINECS: 233-208-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...