4-(Trifluormethylthio)benzaldehyd ≥90%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-((Trifluoromethyl)thio)benzaldehyde
B20313.03
B20313.06
B20313.03EA
49.9
EUR
B20313.03
B20313.06
4-(Trifluormethylthio)benzaldehyd ≥90%
4-(Trifluoromethylthio)benzaldehyde
Purity: 90+%
CAS No.: 4021-50-5
Molecular Formula: C8H5F3OS
Formula Weight: 206.19
Flash Point: 96°(204°F)
Refractive Index: 1.5110
Odour: Stench
Melting Point: 29-31°
Boiling Point: 219°
Beilstein Reference: 2328411
MDL No.: MFCD00129191
Formel:
C₈H₅F₃OS Siedepunkt: 219 °C Schmelzpunkt: 29…31 °C Dichte: 1,310 Flammpunkt: 96 °C (204 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129191 CAS-Nummer: 4021-50-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...