Natrium-1-undecansulfonat, (max. 2% H₂O) 98+% (Trockengewicht)

Lieferant: Thermo Fisher Scientific

B20332.14 B20332.22
B20332.14EA 186 EUR
B20332.14 B20332.22
Natrium-1-undecansulfonat, (max. 2% H₂O) 98+% (Trockengewicht)
Natrium-1-undecansulfonat

Purity: 98+% CAS No.: 5838-34-6 Molecular Formula: CH3(CH2)10SO3Na Formula Weight: 258.36 Melting Point: 300° Beilstein Reference: 3751377

Formel: C₁₂H₂₅NaO₃S
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00053627
CAS-Nummer: 5838-34-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR