(Trifluormethyl)trimethylsilan ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: trimethyl(trifluoromethyl)silane

B20347.06
B20347.06EA 104 EUR
B20347.06
(Trifluormethyl)trimethylsilan ≥98%
(Trifluoromethyl)trimethylsilane

Purity: 98% CAS No.: 81290-20-2 Molecular Formula: C4H9F3Si Formula Weight: 142.20 Flash Point: -10°(14°F) Refractive Index: 1.3305 Boiling Point: 54-55° Beilstein Reference: 4241868 UN No.: 1993 MDL No.: MFCD00145454

Formel: C₄H₉F₃Si
Siedepunkt: 54…55 °C
Dichte: 0,962
Flammpunkt: –10 °C (14 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00145454
CAS-Nummer: 81290-20-2
ADR: 3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR